Learn More
Cayman Chemical PrecoatedGoat ntI-Mouse Ig 5ea
SDP
Catalog No. 502942139
Encompass_Preferred
An ionizable cationic lipid has been used in the generation of lipid nanoparticles for the delivery of mRNA in vitro or in vivo LNPs containing C14-4 and encapsulating mRNA encoding CAR induce CAR expression in primary human T cells ex vivo to the same level as expression induced via electroporation but with reduced cytotoxicity and induce cytotoxicity in Nalm-6 ALL cells in vitro LNPs containing C14-4 and deoxycholic acid and encapsulating mRNA encoding luciferase accumulate in the uterus when intraperitoneally administered and in the lungs and small intestine when intravenously administered in mice
Catalog No. 50-294-2139 Supplier Cayman Chemical Supplier No. 4000095 EA
An ionizable cationic lipid has been used in the generation of lipid nanoparticles for the delivery of mRNA in vitro or in vivo LNPs containing C14-4 and encapsulating mRNA encoding CAR induce CAR expression in primary human T cells ex vivo to the same level as expression induced via electroporation but with reduced cytotoxicity and induce cytotoxicity in Nalm-6 ALL cells in vitro LNPs containing C14-4 and deoxycholic acid and encapsulating mRNA encoding luciferase accumulate in the uterus when intraperitoneally administered and in the lungs and small intestine when intravenously administered in mice