Learn More
Cayman Chemical PrecoatedMouse ntI-RabbIt 5ea
SDP
Catalog No. 502942056
Encompass_Preferred
An ago-PAM of a7 nAChRs enhances ACh-induced activation in a two-electrode voltage-clamp assay using Xenopus oocytes expressing the human a7 nAChR at 10 UM decreases paw edema and induces antinociception in the first and second phase of the formalin test in a mouse model of inflammatory pain at 10 mg/kg inhibits tau aggregation induced by seeding with insoluble human tau isolated from a patient with Alzheimers disease in primary rat neurons
Catalog No. 50-294-2056 Supplier Cayman Chemical Supplier No. 4000045 EA
An ago-PAM of a7 nAChRs enhances ACh-induced activation in a two-electrode voltage-clamp assay using Xenopus oocytes expressing the human a7 nAChR at 10 UM decreases paw edema and induces antinociception in the first and second phase of the formalin test in a mouse model of inflammatory pain at 10 mg/kg inhibits tau aggregation induced by seeding with insoluble human tau isolated from a patient with Alzheimers disease in primary rat neurons