Learn More
Rice Lake Weighing System TP-Series 220gx0.001g Tuning Fork Precision Balance Weight, Unit Count and Percentage Modes, 12 Units of Measure, LCD Display, 115 VAC includes weighing chamber
Catalog No. 501542711
Encompass_Preferred
TP-Series 220gx0.001g Tuning Fork Precision Balance Weight, Unit Count and Percentage Modes, 12 Units of Measure, LCD Display, 115 VAC
Catalog No. 50-154-2711 Supplier Rice Lake Weighing System Supplier No. 107244
TP-Series 220gx0.001g Tuning Fork Precision Balance Weight, Unit Count and Percentage Modes, 12 Units of Measure, LCD Display, 115 VAC