Learn More
Chemplex Industries PRECUT CIRCLES MYLAR 0.10MIL
Supplier:  Chemplex Industries 106
Encompass_Preferred
CHEMPLEX THIN-FILM, PRECUT CIRCLES, Mylar(TM), 2.5u (0.10mil), 2.5 Dia. (6.4cm)
Catalog No. 50-249-334
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.