Learn More
Chemplex Industries PRECUT CIRCLES POLYPROP .24MIL
Catalog No. 50249338
Encompass_Preferred
CHEMPLEX THIN-FILM, PRECUT CIRCLES, Polypropylene, 6.0u (0.24mil), 2.5 Dia. (6.4cm)
Catalog No. 50-249-338 Supplier Chemplex Industries Supplier No. 436
CHEMPLEX THIN-FILM, PRECUT CIRCLES, Polypropylene, 6.0u (0.24mil), 2.5 Dia. (6.4cm)