Learn More
Alzet PEEK Tubing for Use with 2 ml ALZET Osmotic Pumps, Ideal in MRI Studies as a Non-metallic Replacement for the ALZET Flow Moderator
SDP
Catalog No. NC2184994
Encompass
ALZETTM PEEK Tubing, pre-cut and sterilized, is a non-metallic replacement for the flow moderator of an ALZET Osmotic Pump for use in studies involving MRI. PEEK has excellent chemical compatibility and is widely used in medical applications. This item, PEEK Tube for 2 ML ALZET Pumps, is the optimal length and diameter for ALZET Pump Models 2ML1, 2ML2 and 2ML4. Instructions for use can be found at https://www.alzet.com/guide-to-use/mri/
This item is not returnable. View return policy
Catalog No. NC2184994 Supplier Alzet Supplier No. 0002511
ALZETTM PEEK Tubing, pre-cut and sterilized, is a non-metallic replacement for the flow moderator of an ALZET Osmotic Pump for use in studies involving MRI. PEEK has excellent chemical compatibility and is widely used in medical applications. This item, PEEK Tube for 2 ML ALZET Pumps, is the optimal length and diameter for ALZET Pump Models 2ML1, 2ML2 and 2ML4. Instructions for use can be found at https://www.alzet.com/guide-to-use/mri/