Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety™ PredaFlex Shoulder Length Nitrile Glove

Catalog No. 19034394 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-034-394 Large
19-034-389 Small
19-034-393 Medium
19-034-388 X-Large
19-034-387 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-034-394 Supplier MCR Safety™ Supplier No. MG9796L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MCR Safety PredaFlex™ Work Gloves - shoulder length fully coated with nitrile, soft interlock liner, sandy finish.

  • Grommet on the shoulder end for hanging the glove
  • Sandy nitrile finish
  • Shoulder length protection
  • Soft cotton brushed interlock liner
  • Treated with ActiFresh™

Specifications

Accelerator Free No
Applications Industrial
AQL 0.65
Color Yellow
Disposable Reusable
Exterior Finish Smooth
Latex Free No
Material Nitrile
Powdered Powder Free
Product Type Nitrile Gloves
Size Large
Sterility Non-sterile
Thickness (English) 48 mil
Thickness (Metric) 1.21 mm
Glove Cuff Style Elasticated
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.