Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Predator™ Over the Knuckle Nitrile Coated Work Gloves

Catalog No. 191300342C Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Size:
Large
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-130-0342C X-Large
19-041919 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-130-0342C Supplier MCR Safety Supplier No. 9760XL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nitrile-coated gloves feature a cotton jersey lining for comfort, and a safety cuff for increased protection. Acti-Fresh™ treatment helps prevent odors while also eliminating bacteria.

  • Premium nitrile coating on the palm, fingers, and over knuckles
  • Soft jersey lining for increase comfort
  • Safety cuff for additional wrist protection
  • Treated with ActiFresh™ to eliminate odors and bacteria
  • Materials comply with all relevant federal regulations for food contact

Specifications

Certifications/Compliance ANSI A2, FDA
Coated Area Palm, Fingers, Knuckles
Construction Jersey Knit
Coating Material Nitrile
Color Blue
Cut Resistance Rating ANSI A2
Exterior Finish No Texture
Grade Premium
Hand Specific
Material Cotton
Product Type Dip-coated Gloves
Size X-Large
For Use With (Application) Agriculture, Carpentry, Construction, Demolition, Forestry, Landscaping, and Material Handling
Glove Cuff Color Natural
Glove Cuff Style Safety
Liner Material Cotton Jersey
Packaging Type Polybag
Resistant To Abrasion, Contact Heat
Series 9760
Thumb Style Straight
Length (English) 11 in.
Length (Metric) 28 cm
Product Line Predator
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.