Learn More
MCR Safety Predator™ PD2 Safety Glasses
Catalog No. 19812777 Shop All MCR Safety Products
Encompass_Preferred
Change view
Click to view available options
Lens Tint:
Gray
Catalog No. Lens Tint
19-812-777 Gray
1 options
This item is not returnable. View return policy
Catalog No. 19-812-777 Supplier MCR Safety Supplier No. PD1212PF
This item is not returnable. View return policy
MCR Safety Predator™ PD2 Safety Glasses are a favorite for the mining industry and any industry needing dust, impact and anti-fog protection.
  • Meets the ANSI/ISEA Z87.1 D4 designation for dust protection and ANSI/ISEA Z87.1 for high impact
  • Full foam venting helps to reduce heat buildup and assists in minimizing fogging
  • Available in standard anti-fog or MAX6™ superior anti-fog
  • Wide profile lens provides excellent peripheral vision
  • Elastic strap with hook and loop, and 5-point ratchet provide complete adjustability
Specifications
Description Dialectric
Lens Tint Gray
Frame Material Polyurethane