Learn More
Medchemexpress LLC Prednisolone | 50-24-8 | MFCD00003649 | 99.9% | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHY17463
Encompass_Preferred
Prednisolone is a potent, orally active corticosteroid and a glucocorticoid. It exhibits approximately four times the anti-inflammatory activity of hydrocortisone while causing less salt and water retention. Prednisolone is suitable for ocular and anti-inflammatory research purposes.
  • Potent, orally active corticosteroid and glucocorticoid.
  • Exhibits anti-inflammatory activity, approximately four times that of hydrocortisone.
  • Causes less salt and water retention.
  • Suitable for ocular and anti-inflammatory research.
  • Inhibits human leukocyte mitosis and prevents renal disease.
  • Demonstrates agonist activity at human mineralocorticoid receptor.
  • Involved in clinical trials for conditions such as metastatic cancer and and lupus nephritis.
Catalog No. 50-003-85548