Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prednisolone, 99%, Thermo Scientific Chemicals

Catalog No. p-4813626
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC449470010 1 g Glass bottle
AC449470050 5 g Glass bottle
AC449470250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC449470010 Supplier Thermo Scientific Chemicals Supplier No. 449470010
Only null left
Add to Cart

Prednisolone, or metacortandralone, CAS # 50-24-8, is a synthetic glucocorticoid.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• Prednisolone is a derivative of prednisone with an oxo to beta-hydroxy reduction at position 11

• Prednisolone binds cell surface receptors and is transported to the nucleus, where it binds certain nuclear receptors responsible for transcription of proinflammatory cytokines

Applications

• Prednisolone can be used for laboratory research on tumor growth and prevention

• Prednisolone may also be used for developmental and endocrine research, in vivo or in vitro

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50-24-8
Molecular Formula C21H28O5
Molecular Weight (g/mol) 360.44
InChI Key OIGNJSKKLXVSLS-VWUMJDOOSA-N
Synonym prednisolone, metacortandralone, hydroretrocortine, predonine, deltacortril, meticortelone, deltahydrocortisone, prenolone, hydroretrocortin, delta-cortef
PubChem CID 5755
ChEBI CHEBI:8378
IUPAC Name (8S,9S,10R,11S,13S,14S,17R)-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-7,8,9,11,12,14,15,16-octahydro-6H-cyclopenta[a]phenanthren-3-one
SMILES CC12CC(C3C(C1CCC2(C(=O)CO)O)CCC4=CC(=O)C=CC34C)O

Specifications

Melting Point 240.0°C
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15,7841
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in methanol, dioxane, slightly soluble in acetone, chloroform,, ethylacetate
Formula Weight 360.44
Percent Purity 99%
Chemical Name or Material Prednisolone
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May damage the unborn child.
Precautionary Statement GHS P Statement
Use personal protective equipment as required.
Do not handle until all safety precautions have been read and understood.
EINECSNumber 200-021-7

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.