Learn More
Apexbio Technology LLC Prednisolone Acetate 52-21-1 250mg
SDP
Catalog No. 5000093108
Encompass_Preferred
Prednisolone acetate (52-21-1) is a synthetic glucocorticoid derivative targeting glucocorticoid receptors (GR) It is designed to modulate GR-mediated signaling pathways thereby regulating inflammatory and immune response gene expression Prednisolone acetate exerts its biological activity primarily through binding to cytosolic GR forming complexes that translocate into the nucleus to modulate transcriptional events related to inflammation and immune function Based on these pharmacological properties prednisolone acetate holds research potential in the investigation of inflammatory and allergic responses immune regulation and transcriptional modulation in experimental models of chronic inflammation
Catalog No. 50-000-93108 Supplier Apexbio Technology LLC Supplier No. B2013250
Prednisolone acetate (52-21-1) is a synthetic glucocorticoid derivative targeting glucocorticoid receptors (GR) It is designed to modulate GR-mediated signaling pathways thereby regulating inflammatory and immune response gene expression Prednisolone acetate exerts its biological activity primarily through binding to cytosolic GR forming complexes that translocate into the nucleus to modulate transcriptional events related to inflammation and immune function Based on these pharmacological properties prednisolone acetate holds research potential in the investigation of inflammatory and allergic responses immune regulation and transcriptional modulation in experimental models of chronic inflammation