Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prednisolone acetate | 52-21-1 | 99.3% | 1 ML
SDP

Catalog No. 5000411151
Encompass_Preferred

Prednisolone acetate is an adrenocortical hormone active molecule. It exhibits various pharmacological effects, including anti-inflammatory, anti-allergic, and immune suppression activities.

  • Exhibits anti-inflammatory, anti-allergic, and immune suppression activities
  • Targets Glucocorticoid Receptor
  • Involved in immunology/inflammation and vitamin D related/nuclear receptor pathways
  • Prolongs corneal allograft survival in a rat keratoplasty model
  • Soluble in DMSO at 50 mg/mL (124.23 mM), requires ultrasonication
  • Recommended storage in solvent: -80°C for 2 years, -20°C for 1 year
  • For research use only

Catalog No. 50-004-11151 Supplier Medchemexpress LLC Supplier No. HYB121410MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prednisolone acetate is an adrenocortical hormone active molecule. It exhibits various pharmacological effects, including anti-inflammatory, anti-allergic, and immune suppression activities.

  • Exhibits anti-inflammatory, anti-allergic, and immune suppression activities
  • Targets Glucocorticoid Receptor
  • Involved in immunology/inflammation and vitamin D related/nuclear receptor pathways
  • Prolongs corneal allograft survival in a rat keratoplasty model
  • Soluble in DMSO at 50 mg/mL (124.23 mM), requires ultrasonication
  • Recommended storage in solvent: -80°C for 2 years, -20°C for 1 year
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.