Learn More
Selleck Chemical LLC Prednisolone Acetate S2570-50mg
SDP
Catalog No. 503112721
Encompass_Preferred
Prednisolone Acetate (Omnipred Prednisolone-21-acetate) is a synthetic corticosteroid drug that is particularly effective as an immunosuppressant agent
Catalog No. 50-311-2721 Supplier Selleck Chemical LLC Supplier No. S257050MG
Prednisolone Acetate (Omnipred Prednisolone-21-acetate) is a synthetic corticosteroid drug that is particularly effective as an immunosuppressant agent