Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Prednisolone Acetate, Micronized, USP, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18605791 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-791 500 g Amber Glass Bottle
18-605-787 5 g Amber Glass Bottle
18-605-788 25 g Amber Glass Bottle
18-605-789 100 g Amber Glass Bottle
18-605-790 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-605-791 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. PR100500GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

PR100, 52-21-1, C23H30O6

Spectrum™ Chemical Prednisolone Acetate, Micronized, USP is a steroid medication used to treat certain types of allergies, inflammatory conditions, autoimmune disorders, and cancers. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 52-21-1
Molecular Formula C23H30O6
Molecular Weight (g/mol) 402.49
InChI Key LRJOMUJRLNCICJ-JZYPGELDSA-N
IUPAC Name 2-[(1R,3aS,3bS,9aR,9bS,10S,11aS)-1,10-dihydroxy-9a,11a-dimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl acetate
SMILES CC(=O)OCC(=O)[C@@]1(O)CC[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@H]3[C@@H](O)C[C@]12C

Specifications

CAS Max % 1
Loss on Drying 0.01
Packaging Amber Glass Bottle
Quantity 500 g
Specific Rotation +112° to +119°
Percent Purity 97 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Prednisolone Acetate, Micronized
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.