Learn More
Medchemexpress LLC Prednisolone acetate (Prednisolone 21-acetate) | 52-21-1 | 99.3% | 500 MG
SDP
Catalog No. 5000413826
Encompass_Preferred
Prednisolone acetate is an adrenocortical hormone active molecule with various pharmacological effects, including anti-inflammatory, anti-allergic, and immune suppression. It has been shown to prolong corneal allograft survival in a rat keratoplasty model.
  • Administration was via subconjunctival implantation of 40 wt% in microfilm for 12 weeks, or as 1% eye drops applied topically three times daily for 4 weeks.
  • These methods reduced the mean rejection index and prolonged graft survival.
Catalog No. 50-004-13826 Supplier Medchemexpress LLC Supplier No. HYB1214500MG
Prednisolone acetate is an adrenocortical hormone active molecule with various pharmacological effects, including anti-inflammatory, anti-allergic, and immune suppression. It has been shown to prolong corneal allograft survival in a rat keratoplasty model.
  • Administration was via subconjunctival implantation of 40 wt% in microfilm for 12 weeks, or as 1% eye drops applied topically three times daily for 4 weeks.
  • These methods reduced the mean rejection index and prolonged graft survival.