Learn More
Medchemexpress LLC Prednisolone-d7 | 367.49 | 1 MG
SDP
Catalog No. 5000391272
Encompass_Preferred
Prednisolone-d7 is a deuterium-labeled compound of Prednisolone, suitable for research applications. This product requires specific storage conditions to maintain its stability, with powder formulations stable for up to 3 years at -20°C and 2 years at 4°C. When in solvent, it remains stable for 6 months at -80°C and 1 month at -20°C.
  • For research use only
  • Not fully validated for medical applications
  • Store powder at -20°C for up to 3 years
  • Store powder at 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months
  • Store in solvent at -20°C for up to 1 month
Catalog No. 50-003-91272 Supplier Medchemexpress LLC Supplier No. HY165667S1MG
Prednisolone-d7 is a deuterium-labeled compound of Prednisolone, suitable for research applications. This product requires specific storage conditions to maintain its stability, with powder formulations stable for up to 3 years at -20°C and 2 years at 4°C. When in solvent, it remains stable for 6 months at -80°C and 1 month at -20°C.
  • For research use only
  • Not fully validated for medical applications
  • Store powder at -20°C for up to 3 years
  • Store powder at 4°C for up to 2 years
  • Store in solvent at -80°C for up to 6 months
  • Store in solvent at -20°C for up to 1 month