Learn More
Medchemexpress LLC Prednisolone disodium phosphate | 125-02-0 | 99.0%
SDP
Catalog No. 5000408539
Encompass_Preferred
Prednisolone disodium phosphate is a synthetic glucocorticoid that possesses anti-inflammatory and immunomodulating properties. It can be rapidly and completely hydrolyzed to Prednisolone in the plasma.
  • Acts as a synthetic glucocorticoid
  • Possesses anti-inflammatory properties
  • Exhibits immunomodulating properties
  • Rapidly and completely hydrolyzed to Prednisolone in plasma
  • Appears as a white to off-white solid
  • Has a molecular formula of C₂₁H₂₇Na₂O₈P
  • Features a molecular weight of 484.39
  • High purity (typically 99.50%)
Catalog No. 50-004-08539 Supplier Medchemexpress LLC Supplier No. HYB0645R50MG
Prednisolone disodium phosphate is a synthetic glucocorticoid that possesses anti-inflammatory and immunomodulating properties. It can be rapidly and completely hydrolyzed to Prednisolone in the plasma.
  • Acts as a synthetic glucocorticoid
  • Possesses anti-inflammatory properties
  • Exhibits immunomodulating properties
  • Rapidly and completely hydrolyzed to Prednisolone in plasma
  • Appears as a white to off-white solid
  • Has a molecular formula of C₂₁H₂₇Na₂O₈P
  • Features a molecular weight of 484.39
  • High purity (typically 99.50%)