Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prednisolone (disodium phosphate) | 125-02-0 | MFCD00083682 | 99.0% | 484.39 | C21H27Na2O8P | 10 MG
SDP

Catalog No. 5000298964
Encompass_Preferred

Prednisolone disodium phosphate (Standard) is an analytical reference standard of the corticosteroid prednisolone, supplied for research and analytical applications. It is provided as a solid and in ready-to-use solution formats for assay development and method validation.

  • Analytical reference standard for prednisolone assays.
  • Measured purity 98.98% (HPLC) with assay data provided.
  • Molecular formula C21H27Na2O8P, molecular weight 484.39 g/mol.
  • Available in small to bulk pack sizes, including 10 mg solid and 10 mM solution formats.
  • Supplied with datasheet, certificate of analysis, and SDS for traceability.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-98964 Supplier Medchemexpress LLC Supplier No. HYB0645R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prednisolone disodium phosphate (Standard) is an analytical reference standard of the corticosteroid prednisolone, supplied for research and analytical applications. It is provided as a solid and in ready-to-use solution formats for assay development and method validation.

  • Analytical reference standard for prednisolone assays.
  • Measured purity 98.98% (HPLC) with assay data provided.
  • Molecular formula C21H27Na2O8P, molecular weight 484.39 g/mol.
  • Available in small to bulk pack sizes, including 10 mg solid and 10 mM solution formats.
  • Supplied with datasheet, certificate of analysis, and SDS for traceability.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.