Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prednisolone disodium phosphate | 125-02-0 | MFCD00083682 | 99.50% | 25 MG
SDP

Catalog No. 5000450962
Encompass_Preferred

Prednisolone disodium phosphate is a synthetic glucocorticoid with anti-inflammatory and immunomodulating properties. It is rapidly and completely hydrolyzed into prednisolone in the plasma. This product is intended for research use only.

  • Synthetic glucocorticoid
  • Possesses anti-inflammatory properties
  • Has immunomodulating properties
  • Rapidly and completely hydrolyzed into prednisolone in plasma
  • White to off-white solid appearance
  • Molecular weight of 484.39
  • Chemical formula C21H27Na2O8P
  • Recommended storage at 4°C, sealed and away from moisture
  • In solution, can be stored at -80°C for 6 months or -20°C for 1 month

Catalog No. 50-004-50962 Supplier Medchemexpress LLC Supplier No. 25MGHYB0645R
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prednisolone disodium phosphate is a synthetic glucocorticoid with anti-inflammatory and immunomodulating properties. It is rapidly and completely hydrolyzed into prednisolone in the plasma. This product is intended for research use only.

  • Synthetic glucocorticoid
  • Possesses anti-inflammatory properties
  • Has immunomodulating properties
  • Rapidly and completely hydrolyzed into prednisolone in plasma
  • White to off-white solid appearance
  • Molecular weight of 484.39
  • Chemical formula C21H27Na2O8P
  • Recommended storage at 4°C, sealed and away from moisture
  • In solution, can be stored at -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.