Learn More
Medchemexpress LLC HY-U00098 5mg Medchemexpress, Prednisolone Tebutate CAS:7681-14-3 Purity:>98%
SDP
Catalog No. 501970622
Encompass_Preferred
Medchemexpress, HY-U00098 5mg Prednisolone Tebutate CAS:7681-14-3 Prednisolone tebutate is a synthetic glucocorticoid used as an antiinflammatory and immunosuppressant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0622 Supplier Medchemexpress LLC Supplier No. HYU000985MG
Medchemexpress, HY-U00098 5mg Prednisolone Tebutate CAS:7681-14-3 Prednisolone tebutate is a synthetic glucocorticoid used as an antiinflammatory and immunosuppressant. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.