Learn More
Apexbio Technology LLC Prednisone 53-03-2 1g
SDP
Catalog No. 5000062043
Encompass_Preferred
Prednisone is a synthetic glucocorticoid which functions primarily as an agonist of the glucocorticoid receptor (GR) Upon binding to the GR prednisone induces receptor translocation into the nucleus initiating the regulation of gene expression involved in inflammation immune responses and cellular proliferation Due to its immunosuppressive mechanism prednisone is widely used as an experimental tool in biomedical studies exploring inflammatory diseases autoimmune conditions and immune signaling pathways Structurally prednisone is a corticosteroid derivative characterized by specific modifications that affect its pharmacodynamics and bioavailability In laboratory settings prednisone assists researchers in dissecting glucocorticoid-mediated physiological and molecular events evaluating receptor-ligand interactions and examining downstream signaling mechanisms in various cell-based assays and animal models
Catalog No. 50-000-62043 Supplier Apexbio Technology LLC Supplier No. B21481000
 Restricted Item
Add to Cart
Add to Cart
Prednisone is a synthetic glucocorticoid which functions primarily as an agonist of the glucocorticoid receptor (GR) Upon binding to the GR prednisone induces receptor translocation into the nucleus initiating the regulation of gene expression involved in inflammation immune responses and cellular proliferation Due to its immunosuppressive mechanism prednisone is widely used as an experimental tool in biomedical studies exploring inflammatory diseases autoimmune conditions and immune signaling pathways Structurally prednisone is a corticosteroid derivative characterized by specific modifications that affect its pharmacodynamics and bioavailability In laboratory settings prednisone assists researchers in dissecting glucocorticoid-mediated physiological and molecular events evaluating receptor-ligand interactions and examining downstream signaling mechanisms in various cell-based assays and animal models