Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prednisone | 53-03-2 | 1 G
SDP

Supplier:  Medchemexpress LLC HYB02141G

Encompass_Preferred

Prednisone (Adasone) is a corticosteroid agent with anti-inflammatory and immunosuppressive effects. It can be used to study diseases related to systemic lupus erythematosus.

  • Targets glucocorticoid receptor
  • Involved in apoptosis
  • Used in immunology/inflammation pathway research
  • Related to vitamin D and nuclear receptor pathways
  • Applied in research for cardiovascular disease, inflammation/immunology, endocrinology, and cancer
  • Appears as a solid
  • Melting/freezing point: 236-238 °C
  • Soluble in DMSO (125 mg/mL) and H2O (1 mg/mL)
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 1 year or -20°C for 6 months
  • Shipping at room temperature is acceptable if less than 2 weeks

Catalog No. 50-004-05484


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.