Learn More
Medchemexpress LLC Prednisone | 53-03-2 | 99.8% | 25 MG
SDP
Catalog No. 5000450601
Encompass_Preferred
Prednisone is an analytical standard and a corticosteroid agent with anti-inflammatory and immunosuppressive effects. It is intended for research and analytical applications, particularly for studying diseases like systemic lupus erythematosus. This reference standard is suitable for qualitative, quantitative, and methodological research experiments using techniques such as HPLC, GC, and MS.
  • Analytical standard for research
  • Corticosteroid agent with anti-inflammatory effects
  • Immunosuppressive properties
  • Useful for studying systemic lupus erythematosus
  • Compatible with HPLC, GC, and MS research experiments
Catalog No. 50-004-50601 Supplier Medchemexpress LLC Supplier No. 25MGHYB0214R
Prednisone is an analytical standard and a corticosteroid agent with anti-inflammatory and immunosuppressive effects. It is intended for research and analytical applications, particularly for studying diseases like systemic lupus erythematosus. This reference standard is suitable for qualitative, quantitative, and methodological research experiments using techniques such as HPLC, GC, and MS.
  • Analytical standard for research
  • Corticosteroid agent with anti-inflammatory effects
  • Immunosuppressive properties
  • Useful for studying systemic lupus erythematosus
  • Compatible with HPLC, GC, and MS research experiments