Learn More
Selleck Chemical LLC Prednisone acetate
SDP
Catalog No. 5000003185
Encompass_Preferred
Prednisone acetate (Betapar Cortan Deltasone Fernisone Meticorten Prednisone 21-acetate) is a glucocorticoid steroid that can be used as a glucocorticoid receptor agonist with anti-inflammatory and immunomodulating properties
Catalog No. 50-000-03185 Supplier Selleck Chemical LLC Supplier No. S5489100MG
Prednisone acetate (Betapar Cortan Deltasone Fernisone Meticorten Prednisone 21-acetate) is a glucocorticoid steroid that can be used as a glucocorticoid receptor agonist with anti-inflammatory and immunomodulating properties