Learn More
Medchemexpress LLC Prednisone acetate-d3 | 125-10-0 | 96.5% | 403.48 | C23H25D3O6 | 10 MG
SDP
Catalog No. 5000298056
Encompass_Preferred
Prednisone acetate-d3 is the deuterium-labeled analog of prednisone 21-acetate used as a stable isotope internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It serves as a tracer in pharmacokinetic and metabolic studies and is supplied as a solid powder with storage recommendations for long-term stability.
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • Suitable tracer for pharmacokinetic and metabolic studies.
  • High chemical purity (~96.5%) to support accurate quantitation.
  • Solid powder form with recommended low-temperature storage for stability.
Catalog No. 50-002-98056 Supplier Medchemexpress LLC Supplier No. HYB1832S10MG
Prednisone acetate-d3 is the deuterium-labeled analog of prednisone 21-acetate used as a stable isotope internal standard for quantitative analysis by NMR, GC-MS, and LC-MS. It serves as a tracer in pharmacokinetic and metabolic studies and is supplied as a solid powder with storage recommendations for long-term stability.
  • Deuterium-labeled internal standard for NMR, GC-MS, and LC-MS.
  • Suitable tracer for pharmacokinetic and metabolic studies.
  • High chemical purity (~96.5%) to support accurate quantitation.
  • Solid powder form with recommended low-temperature storage for stability.