Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Prednisone-d8 | 98.0% | 250 UG
SDP

Catalog No. 5000406167
Encompass_Preferred

Prednisone-d8 is a deuterium labeled Prednisone, which is a synthetic corticosteroid and effective immunosuppressant. It is suitable for research use as a tracer and internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS.

  • Molecular weight: 366.48
  • Formula: C21H18D8O5
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 98.00%
  • Ships at room temperature in continental US
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-004-06167 Supplier Medchemexpress LLC Supplier No. 250UGHYB0214S
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prednisone-d8 is a deuterium labeled Prednisone, which is a synthetic corticosteroid and effective immunosuppressant. It is suitable for research use as a tracer and internal standard in quantitative analysis methods such as NMR, GC-MS, or LC-MS.

  • Molecular weight: 366.48
  • Formula: C21H18D8O5
  • Appearance: Solid
  • Color: White to off-white
  • Purity: 98.00%
  • Ships at room temperature in continental US
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.