Learn More
Medchemexpress LLC Prednisone (Standard) | 53-03-2 | MFCD00003608 | 99.8% | 100 MG
SDP
Catalog No. 5000406137
Encompass_Preferred
Prednisone (Standard), also known as Dehydrocortisone (Standard), is an analytical standard of Prednisone. This corticosteroid agent exhibits anti-inflammatory and immunosuppressive effects and can be utilized to study diseases such as systemic lupus erythematosus. It is intended for research and analytical applications, commonly employed in qualitative, quantitative, and methodological experiments using techniques like HPLC, GC, and MS. Its molecular formula is C₂₁H₂₆O₅, with a molecular weight of 358.43, and it appears as a white to off-white solid.
  • Analytical standard for research and analytical applications.
  • High purity (99.81%) for reliable results.
  • Useful in studying systemic lupus erythematosus.
  • Supports various analytical methods like HPLC, GC, and MS.
Catalog No. 50-004-06137 Supplier Medchemexpress LLC Supplier No. HYB0214R100MG
Prednisone (Standard), also known as Dehydrocortisone (Standard), is an analytical standard of Prednisone. This corticosteroid agent exhibits anti-inflammatory and immunosuppressive effects and can be utilized to study diseases such as systemic lupus erythematosus. It is intended for research and analytical applications, commonly employed in qualitative, quantitative, and methodological experiments using techniques like HPLC, GC, and MS. Its molecular formula is C₂₁H₂₆O₅, with a molecular weight of 358.43, and it appears as a white to off-white solid.
  • Analytical standard for research and analytical applications.
  • High purity (99.81%) for reliable results.
  • Useful in studying systemic lupus erythematosus.
  • Supports various analytical methods like HPLC, GC, and MS.