Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ PREempt™ Plus
GREENER_CHOICE

Catalog No. 19084210 Shop All Contec Inc. Products
Change view
Click to view available options
Volume (Metric):
0.9 L
3.8 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
19-084-210 0.9 L
19-084-211 3.8 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-084-210 Supplier Contec™ Supplier No. 2B101
Add to Cart
Add to Cart

New and improved formulation of PREempt™ RTU that features faster kill times and lower residue

  • Utilizes Accelerated Hydrogen Peroxide™ (AHP™), a patented synergistic blend of commonly used ingredients, when combined with low levels of hydrogen peroxide dramatically increase its efficacy and cleaning performance
  • Ideal for cleaning and disinfecting environmental surfaces in cleanrooms and laboratory areas including workstations, fume hoods, equipment and other hard non-porous environmental surfaces
  • Ready-to-use, one step broad-spectrum cleaner and disinfectant designed for cleaning and disinfecting hard, non-porous surfaces
  • Kills viruses in 30 sec., bacteria in 1 min., fungi in 30 sec. and TB in 1 min.
  • Lower residue than original PREempt™ RTU making it more suitable for critical environments
  • Contains no volatile organic compounds
  • Health hazard rating of zero

Specifications

Product Type Cleaner and Disinfectant Solution
pH 2 to 2.5
Sterility Non-sterile
Composition Hydrogen Peroxide
For Use With (Application) Cleaning and disinfecting environmental surfaces in cleanrooms and labs
Volume (English) 32 oz.
Volume (Metric) 0.9 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.