Learn More
Fisherbrand™ Fisherbrand Prefilled 2 ml Bead Tubes for Refrigerated Bead Homogenizers
Catalog No. 14828342 Shop All Fisherbrand Products
Change view
Click to view available options
Particle Size:
0.1 mm
0.5 mm
1.5 mm
2.8 mm
3 mm
6 mm
Various
Material:
Ceramic, Silica
Garnet, Ceria-based Ceramic
Silica
Stainless Steel
Zirconium
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size Material
14-828-342 Various Ceramic, Silica
14-828-341 6 mm Garnet, Ceria-based Ceramic
14-828-348 2.8 mm Stainless Steel
14-828-347 3 mm Zirconium
14-828-346 1.5 mm Zirconium
14-828-345 0.5 mm Zirconium
14-828-344 0.5 mm Silica
14-828-343 0.1 mm Silica
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14-828-342
Add to Cart
Add to Cart
2 ml bead tubes are ready-to-use and available with a variety of bead types/sizes for homogenization of a wide range of sample types
  • 2 ml tubes prefilled with an appropriate amount of beads
  • Range of bead material/size for a variety of sample types
  • Ready-to-use
  • Zirconium beads are certified nuclease and protease free
  • High quality screw capped tubes with o-ring
  • Supplied in packs of 50
Specifications
Product Type Prefilled Bead Homogenizer Tubes
Particle Size Various
Autoclavable Autoclavable
Material Ceramic, Silica
Closure Color Clear
For Use With (Application) Homogenizations of Soil, Sludge, Feces and Environmental Samples, Acid Wahed And Heat Treatment
For Use With (Equipment) Bead Homogenizers
Packaging Quantity 50 Pk.