Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Fisherbrand Prefilled 2 ml Bead Tubes for Refrigerated Bead Homogenizers

Catalog No. 14828348 Shop All Fisherbrand Products
Change view
Click to view available options
Particle Size:
0.1 mm
0.5 mm
1.5 mm
2.8 mm
3 mm
6 mm
Various
Material:
Ceramic, Silica
Garnet, Ceria-based Ceramic
Silica
Stainless Steel
Zirconium
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Particle Size Material
14-828-348 2.8 mm Stainless Steel
14-828-342 Various Ceramic, Silica
14-828-341 6 mm Garnet, Ceria-based Ceramic
14-828-347 3 mm Zirconium
14-828-346 1.5 mm Zirconium
14-828-345 0.5 mm Zirconium
14-828-344 0.5 mm Silica
14-828-343 0.1 mm Silica
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 14-828-348
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2 ml bead tubes are ready-to-use and available with a variety of bead types/sizes for homogenization of a wide range of sample types

  • 2 ml tubes prefilled with an appropriate amount of beads
  • Range of bead material/size for a variety of sample types
  • Ready-to-use
  • Zirconium beads are certified nuclease and protease free
  • High quality screw capped tubes with o-ring
  • Supplied in packs of 50

Specifications

Particle Size 2.8 mm
Material Stainless Steel
Closure Color Clear
For Use With (Application) Tough and Hard Samples, Alcohol Washed and Heat treatment
For Use With (Equipment) Bead Homogenizers
Product Type Prefilled Bead Homogenizer Tubes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.