Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger RAE Pavement Pre-Determined Message, Exit Legend, Thermoplastic, 5-1/8 ft x 8 ft, 90 mil, 1 ea

Catalog No. 502778103
Encompass_Preferred

The Preformed Thermoplastic EXIT marking is designed for high-visibility pavement messaging. This durable thermoplastic material provides long-lasting performance for traffic safety applications and is engineered for ease of installation using a hand torch.

  • Legend: Exit
  • Material: Thermoplastic
  • Thickness: 90 mil
  • Overall dimensions: 8 ft (length) x 5-1/8 ft (width)
  • Application method: Hand torch
  • Color: White
  • Compliant with ASTM D 6628, ASTM E 303, MUTCD, FHWA, and AASHTO M-249 standards
  • Shelf life: 2 yr

Catalog No. 50-277-8103 Supplier Grainger Supplier No. 18E519
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

The Preformed Thermoplastic EXIT marking is designed for high-visibility pavement messaging. This durable thermoplastic material provides long-lasting performance for traffic safety applications and is engineered for ease of installation using a hand torch.

  • Legend: Exit
  • Material: Thermoplastic
  • Thickness: 90 mil
  • Overall dimensions: 8 ft (length) x 5-1/8 ft (width)
  • Application method: Hand torch
  • Color: White
  • Compliant with ASTM D 6628, ASTM E 303, MUTCD, FHWA, and AASHTO M-249 standards
  • Shelf life: 2 yr

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.