Learn More
Grainger RAE Products Preformed Thermoplastic Pavement Markings, Lines, White, 125 Mil, 4 in. x 30 ft., 3/Pk.
Catalog No. 502784893
Encompass_Preferred
These white preformed thermoplastic pavement marking rolls provide a durable and high-visibility solution for line striping and zone marking on exterior asphalt or concrete surfaces. Engineered for efficiency, these markings can be applied using a standard propane torch, eliminating the need for specialized equipment.
  • Designed for high-visibility night-time reflectivity with integrated glass bead technology.
  • Provides 30 sq. ft. of coverage per pack with three rolls measuring 4 in. wide by 30 ft. long.
  • Features long-term wear resistance suitable for high-traffic areas.
  • Bonds strongly to pavement surfaces with a fast 2 min. dry time.
  • Meets multiple industry standards including AASHTO M-249 and ASTM D 4592.
  • Zero VOC content for environmentally conscious applications.
Catalog No. 50-278-4893 Supplier Grainger Supplier No. 18E646
These white preformed thermoplastic pavement marking rolls provide a durable and high-visibility solution for line striping and zone marking on exterior asphalt or concrete surfaces. Engineered for efficiency, these markings can be applied using a standard propane torch, eliminating the need for specialized equipment.
  • Designed for high-visibility night-time reflectivity with integrated glass bead technology.
  • Provides 30 sq. ft. of coverage per pack with three rolls measuring 4 in. wide by 30 ft. long.
  • Features long-term wear resistance suitable for high-traffic areas.
  • Bonds strongly to pavement surfaces with a fast 2 min. dry time.
  • Meets multiple industry standards including AASHTO M-249 and ASTM D 4592.
  • Zero VOC content for environmentally conscious applications.