Learn More
Grainger RAE Preformed Thermoplastic Pavement Markings, White, 125 mil, 15/Pk.
Catalog No. 502784595
Encompass_Preferred
These preformed thermoplastic pavement markings provide a durable solution for traffic lane delineation, combining the convenience of pre-cut materials with the performance characteristics of hot-applied thermoplastic. Designed for exterior pavement use, these white lines arrive ready for installation, ensuring efficient application and immediate results.
  • Embedded with high-quality reflective glass beads for enhanced visibility
  • Skid-resistant surface for improved safety
  • Efficient installation using a standard hand torch
  • Suitable for a wide temperature range of 32 °F to 275 °F
  • Durable 125 mil thickness
  • Certified to AASHTO M-249, AASHTO T-250, ASTM D 4592, ASTM D 6628, ASTM E 1710, and ASTM E 303 standards
  • Fast dry time of 2 min
Catalog No. 50-278-4595 Supplier Grainger Supplier No. 18E650
These preformed thermoplastic pavement markings provide a durable solution for traffic lane delineation, combining the convenience of pre-cut materials with the performance characteristics of hot-applied thermoplastic. Designed for exterior pavement use, these white lines arrive ready for installation, ensuring efficient application and immediate results.
  • Embedded with high-quality reflective glass beads for enhanced visibility
  • Skid-resistant surface for improved safety
  • Efficient installation using a standard hand torch
  • Suitable for a wide temperature range of 32 °F to 275 °F
  • Durable 125 mil thickness
  • Certified to AASHTO M-249, AASHTO T-250, ASTM D 4592, ASTM D 6628, ASTM E 1710, and ASTM E 303 standards
  • Fast dry time of 2 min