Learn More
Grainger RAE Preformed Thermoplastic Pavement Markings, Yellow, 6 in. Length, 3 ft. Width, 32/Pk.
Catalog No. 502784627
Encompass_Preferred
These heat-activated, preformed thermoplastic pavement markings provide durable, clear messaging for asphalt and concrete surfaces. Designed for exterior applications, these markings are ideal for designating lanes, parking spots, walkways, and rights-of-way. They are flush with the pavement surface, skid resistant, and engineered to withstand heavy use while remaining highly visible.
  • Features thermoplastic construction with 90 mil thickness for long-lasting performance
  • Skid resistant surface for enhanced safety in exterior environments
  • Application is completed via hand torch with a quick 2 min dry time
  • Suitable for application between 32°F and 275°F
  • Complies with various AASHTO and ASTM standards for quality and durability
  • Provides clear visibility with a non-transparent yellow finish
Catalog No. 50-278-4627 Supplier Grainger Supplier No. 18E635
These heat-activated, preformed thermoplastic pavement markings provide durable, clear messaging for asphalt and concrete surfaces. Designed for exterior applications, these markings are ideal for designating lanes, parking spots, walkways, and rights-of-way. They are flush with the pavement surface, skid resistant, and engineered to withstand heavy use while remaining highly visible.
  • Features thermoplastic construction with 90 mil thickness for long-lasting performance
  • Skid resistant surface for enhanced safety in exterior environments
  • Application is completed via hand torch with a quick 2 min dry time
  • Suitable for application between 32°F and 275°F
  • Complies with various AASHTO and ASTM standards for quality and durability
  • Provides clear visibility with a non-transparent yellow finish