Learn More
Grainger RAE Preformed Thermoplastic Pavement Markings, Lines, White, 90 mil, 18 in. Lg, 8/Pk.
Catalog No. 502784854
Encompass_Preferred
These preformed thermoplastic pavement markings provide a durable and high-visibility solution for traffic control and road safety. Engineered for exterior use on pavement, these 90 mil thick white lines feature embedded glass beads from Swarco for enhanced reflectivity in various lighting and weather conditions. The markings are designed for efficient installation using only a propane hand torch, eliminating the need for specialized melting equipment or metal stencils.
  • Durable thermoplastic construction with 90 mil thickness
  • Reflective glass beads embedded for high visibility
  • Simple application using a standard hand torch
  • Rapid dry time of 2 min
  • Compatible with diverse weather conditions
  • Skid resistant surface
  • Meets AASHTO and ASTM standards
Catalog No. 50-278-4854 Supplier Grainger Supplier No. 18E642
These preformed thermoplastic pavement markings provide a durable and high-visibility solution for traffic control and road safety. Engineered for exterior use on pavement, these 90 mil thick white lines feature embedded glass beads from Swarco for enhanced reflectivity in various lighting and weather conditions. The markings are designed for efficient installation using only a propane hand torch, eliminating the need for specialized melting equipment or metal stencils.
  • Durable thermoplastic construction with 90 mil thickness
  • Reflective glass beads embedded for high visibility
  • Simple application using a standard hand torch
  • Rapid dry time of 2 min
  • Compatible with diverse weather conditions
  • Skid resistant surface
  • Meets AASHTO and ASTM standards