Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Rae Pavement Pre-Determined Messages, Thermoplastic, 24 in. x 3 ft., White, 8/Pk.

Catalog No. 502784875
Encompass_Preferred

These preformed thermoplastic pavement markings provide a durable solution for high-visibility traffic signaling and lane delineation on exterior pavement surfaces. Designed for reliable performance and safety, this white thermoplastic material is skid-resistant and features a non-transparent finish to ensure clarity.

  • Features a thickness of 90 mil
  • Applied using a hand torch
  • Suitable for exterior use with a dry time of 2 min
  • Meets AASHTO M-249, AASHTO T-250, ASTM D 4592, ASTM D 6628, ASTM E 1710, and ASTM E 303 standards
  • Application temperature range of 32°F to 275°F
  • Shelf life of 2 yr

Catalog No. 50-278-4875 Supplier Grainger Supplier No. 18E644
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These preformed thermoplastic pavement markings provide a durable solution for high-visibility traffic signaling and lane delineation on exterior pavement surfaces. Designed for reliable performance and safety, this white thermoplastic material is skid-resistant and features a non-transparent finish to ensure clarity.

  • Features a thickness of 90 mil
  • Applied using a hand torch
  • Suitable for exterior use with a dry time of 2 min
  • Meets AASHTO M-249, AASHTO T-250, ASTM D 4592, ASTM D 6628, ASTM E 1710, and ASTM E 303 standards
  • Application temperature range of 32°F to 275°F
  • Shelf life of 2 yr

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.