Learn More
Grainger Rae Pavement Pre-Determined Messages, Thermoplastic, 8 in Length, 3 ft Width, 24/Pk.
Catalog No. 502784895
Encompass_Preferred
These preformed thermoplastic pavement markings provide a durable and highly visible solution for exterior pavement marking needs. Engineered for long-term performance, these markings utilize the properties of hot-applied thermoplastic and incorporate reflective glass beads to ensure visibility.
  • Features a thickness of 90 mil
  • Applied using a hand torch or propane torch
  • Compliant with AASHTO M-249, AASHTO T-250, ASTM D 4592, ASTM D 6628, ASTM E 1710, and ASTM E 303 standards
  • Skid resistant for added safety
  • Dry time of 2 min
  • Compatible with application temperatures between 32°F and 275°F
  • Shelf life of 2 yr
Catalog No. 50-278-4895 Supplier Grainger Supplier No. 18E637
These preformed thermoplastic pavement markings provide a durable and highly visible solution for exterior pavement marking needs. Engineered for long-term performance, these markings utilize the properties of hot-applied thermoplastic and incorporate reflective glass beads to ensure visibility.
  • Features a thickness of 90 mil
  • Applied using a hand torch or propane torch
  • Compliant with AASHTO M-249, AASHTO T-250, ASTM D 4592, ASTM D 6628, ASTM E 1710, and ASTM E 303 standards
  • Skid resistant for added safety
  • Dry time of 2 min
  • Compatible with application temperatures between 32°F and 275°F
  • Shelf life of 2 yr