Learn More
Grainger RAE Preformed Thermoplastic Lines, White, 24 in. Wide, 24 ft. Length, 1 ea.
Catalog No. 502784639
Encompass_Preferred
These preformed thermoplastic lines offer a durable solution for pavement marking, designed for long-term performance on asphalt surfaces. The material is supplied as ready-to-apply pieces that are activated by heat for straightforward field installation.
  • Provides durable long-term performance with an economical investment
  • Lasts 4 to 5 times longer than standard traffic paint
  • Incorporates premium glass beads to provide reflectivity throughout the life of the line
  • Contains zero VOC and heavy metal free pigments for an environmentally friendly design
  • Installs easily with a propane torch at temperatures down to 32 °F
  • Ready for traffic within 5 to 10 minutes after cooling
Catalog No. 50-278-4639 Supplier Grainger Supplier No. 18E658
These preformed thermoplastic lines offer a durable solution for pavement marking, designed for long-term performance on asphalt surfaces. The material is supplied as ready-to-apply pieces that are activated by heat for straightforward field installation.
  • Provides durable long-term performance with an economical investment
  • Lasts 4 to 5 times longer than standard traffic paint
  • Incorporates premium glass beads to provide reflectivity throughout the life of the line
  • Contains zero VOC and heavy metal free pigments for an environmentally friendly design
  • Installs easily with a propane torch at temperatures down to 32 °F
  • Ready for traffic within 5 to 10 minutes after cooling