Learn More
Grainger RAE Preformed Thermoplastic Pavement Markings, Lines, Yellow, 16 in. Lg., 3 ft. Wd., 8/Pk.
Catalog No. 502784695
Encompass_Preferred
These preformed thermoplastic pavement markings are heat-activated, pre-cut lines designed for durable traffic control on asphalt and concrete surfaces. The material provides a flush, skid-resistant finish suitable for exterior walkways, roadways, and parking areas.
  • Features reflective glass beads embedded throughout for high visibility
  • Applied directly to pavement using a standard hand torch
  • Suitable for exterior use on asphalt and concrete
  • Complies with AASHTO M-249 and various ASTM performance standards
  • Thickness of 90 mil
  • Dry time of 2 min
  • Minimum application temperature of 32 °F
  • Maximum application temperature of 275 °F
  • Contains 0 g/L VOC content
Catalog No. 50-278-4695 Supplier Grainger Supplier No. 18E641
These preformed thermoplastic pavement markings are heat-activated, pre-cut lines designed for durable traffic control on asphalt and concrete surfaces. The material provides a flush, skid-resistant finish suitable for exterior walkways, roadways, and parking areas.
  • Features reflective glass beads embedded throughout for high visibility
  • Applied directly to pavement using a standard hand torch
  • Suitable for exterior use on asphalt and concrete
  • Complies with AASHTO M-249 and various ASTM performance standards
  • Thickness of 90 mil
  • Dry time of 2 min
  • Minimum application temperature of 32 °F
  • Maximum application temperature of 275 °F
  • Contains 0 g/L VOC content