Learn More
Grainger Rae Preformed Thermoplastic Pavement Markings, Lines, White, 16/Pk.
Catalog No. 502784750
Encompass_Preferred
These preformed thermoplastic pavement markings provide a durable and highly visible solution for road signage and traffic flow management. Engineered for exterior pavement applications, the material combines the convenience of pre-cut sections with the performance characteristics of hot-applied thermoplastic. The markings feature integrated premium glass beads for enhanced reflectivity and safety in diverse lighting and weather conditions, with an installation process that requires only a propane torch.
  • Features 90 mil thickness for high durability
  • Reflective glass bead integration for day and night visibility
  • Easy application using a standard hand torch
  • Skid-resistant surface for improved road safety
  • Fast installation with a 2 min dry time
  • Suitable for exterior use on pavement surfaces
Catalog No. 50-278-4750 Supplier Grainger Supplier No. 18E638
These preformed thermoplastic pavement markings provide a durable and highly visible solution for road signage and traffic flow management. Engineered for exterior pavement applications, the material combines the convenience of pre-cut sections with the performance characteristics of hot-applied thermoplastic. The markings feature integrated premium glass beads for enhanced reflectivity and safety in diverse lighting and weather conditions, with an installation process that requires only a propane torch.
  • Features 90 mil thickness for high durability
  • Reflective glass bead integration for day and night visibility
  • Easy application using a standard hand torch
  • Skid-resistant surface for improved road safety
  • Fast installation with a 2 min dry time
  • Suitable for exterior use on pavement surfaces