Learn More
Grainger RAE Preformed Thermoplastic Pavement Marking, Only, White, 4/Pk.
Catalog No. 502784635
Encompass_Preferred
These heat-activated, preformed thermoplastic pavement markings provide clear, repeatable messaging for roadways, walkways, and parking areas. Designed for durability and performance, these messages are installed flush to the surface and are suitable for use on asphalt and concrete.
  • Provides enhanced reflectivity using premium glass beads for improved visibility
  • Features skid-resistant properties for safety
  • Designed for exterior application with a dry time of 2 min
  • Compatible with standard hand torch installation methods
  • Complies with industry standards including AASHTO M-249 and ASTM D 4592
  • Dimensions measure 4 ft in length and 34 in in width
Catalog No. 50-278-4635 Supplier Grainger Supplier No. 18E577
These heat-activated, preformed thermoplastic pavement markings provide clear, repeatable messaging for roadways, walkways, and parking areas. Designed for durability and performance, these messages are installed flush to the surface and are suitable for use on asphalt and concrete.
  • Provides enhanced reflectivity using premium glass beads for improved visibility
  • Features skid-resistant properties for safety
  • Designed for exterior application with a dry time of 2 min
  • Compatible with standard hand torch installation methods
  • Complies with industry standards including AASHTO M-249 and ASTM D 4592
  • Dimensions measure 4 ft in length and 34 in in width