Learn More
Grainger RAE Preformed Thermoplastic Pavement Marking, Right, White, 8 ft L, 6 3/4 ft W, 1 ea
Catalog No. 502778102
Encompass_Preferred
These preformed thermoplastic pavement markings provide a durable and highly visible solution for road surface designation. Designed for exterior use, this right-facing marking is constructed from thermoplastic material with embedded premium glass beads to ensure long-lasting performance and visibility. The system is compliant with major regulatory standards and is suitable for installation via hand torch.
  • High-visibility design with reflective glass beads
  • Durable performance for exterior pavement applications
  • Skid resistant surface for enhanced safety
  • Meets ASTM D 6628, ASTM E 303, MUTCD, FHWA, and AASHTO M-249 standards
  • Installation requires a propane torch for adhesion
  • Dimensions: 8 ft long by 6 3/4 ft wide
  • Thickness of 90 mil
Catalog No. 50-277-8102 Supplier Grainger Supplier No. 18E528
These preformed thermoplastic pavement markings provide a durable and highly visible solution for road surface designation. Designed for exterior use, this right-facing marking is constructed from thermoplastic material with embedded premium glass beads to ensure long-lasting performance and visibility. The system is compliant with major regulatory standards and is suitable for installation via hand torch.
  • High-visibility design with reflective glass beads
  • Durable performance for exterior pavement applications
  • Skid resistant surface for enhanced safety
  • Meets ASTM D 6628, ASTM E 303, MUTCD, FHWA, and AASHTO M-249 standards
  • Installation requires a propane torch for adhesion
  • Dimensions: 8 ft long by 6 3/4 ft wide
  • Thickness of 90 mil