Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Rae Pavement Line Rolls, Thermoplastic, 6 in. Width, 30 ft. Length, 2/Pk.

Catalog No. 502784705
Encompass_Preferred

These preformed thermoplastic pavement markings provide durable and skid-resistant lines for exterior surfaces. Designed for application via hand torch, these yellow thermoplastic rolls are engineered for high visibility and long-term performance on pavement.

  • Thickness of 90 mil
  • Overall length of 30 ft
  • Overall width of 6 in
  • Application temperature range of 32°F to 275°F
  • Dry time of 2 min
  • Includes 2 pieces per package
  • Complies with AASHTO M-249, AASHTO T-250, ASTM D 4592, ASTM D 6628, ASTM E 1710, and ASTM E 303 standards
  • Shelf life of 2 yr

Catalog No. 50-278-4705 Supplier Grainger Supplier No. 18E667
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These preformed thermoplastic pavement markings provide durable and skid-resistant lines for exterior surfaces. Designed for application via hand torch, these yellow thermoplastic rolls are engineered for high visibility and long-term performance on pavement.

  • Thickness of 90 mil
  • Overall length of 30 ft
  • Overall width of 6 in
  • Application temperature range of 32°F to 275°F
  • Dry time of 2 min
  • Includes 2 pieces per package
  • Complies with AASHTO M-249, AASHTO T-250, ASTM D 4592, ASTM D 6628, ASTM E 1710, and ASTM E 303 standards
  • Shelf life of 2 yr

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.