Learn More
Grainger Rae Preformed Thermoplastic Pavement Markings, White, 4 in, 30 ft, 3/Pk.
Catalog No. 502784754
Encompass_Preferred
These preformed thermoplastic pavement markings provide a durable and highly visible solution for road and surface navigation. Designed for exterior use, this material features embedded glass beads to ensure enhanced reflectivity and safety under various lighting and weather conditions. The markings are suitable for application on asphalt surfaces, while a primer is recommended for use on concrete or older surfaces.
  • Provides high-visibility markings for traffic flow and navigation
  • Embedded glass beads offer enhanced reflectivity
  • Installs directly to pavement using a simple hand torch
  • Ready for traffic in 5 to 10 minutes
  • Durable performance equivalent to hot-applied thermoplastic materials
  • Meets AASHTO and ASTM standards for quality and performance
Catalog No. 50-278-4754 Supplier Grainger Supplier No. 18E660
These preformed thermoplastic pavement markings provide a durable and highly visible solution for road and surface navigation. Designed for exterior use, this material features embedded glass beads to ensure enhanced reflectivity and safety under various lighting and weather conditions. The markings are suitable for application on asphalt surfaces, while a primer is recommended for use on concrete or older surfaces.
  • Provides high-visibility markings for traffic flow and navigation
  • Embedded glass beads offer enhanced reflectivity
  • Installs directly to pavement using a simple hand torch
  • Ready for traffic in 5 to 10 minutes
  • Durable performance equivalent to hot-applied thermoplastic materials
  • Meets AASHTO and ASTM standards for quality and performance