Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grainger Rae Pavement Line Rolls, Thermoplastic, White, 30 ft, 16 in., 1 ea

Catalog No. 502784756
Encompass_Preferred

These preformed thermoplastic pavement markings are designed for use as durable legend rolls on exterior pavement surfaces. The material provides a non-transparent, skid-resistant finish in white, suitable for professional traffic marking applications.

  • Meets AASHTO M-249, AASHTO T-250, ASTM D 4592, ASTM D 6628, ASTM E 1710, and ASTM E 303 standards
  • Application method requires a hand torch with an application temperature range of 32°F to 275°F
  • Durable construction with 90 mil thickness
  • Fast dry time of 2 min
  • Shelf life of 2 yr
  • Dimensions of 30 ft Lg by 16 in Wd

Catalog No. 50-278-4756 Supplier Grainger Supplier No. 18E670
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These preformed thermoplastic pavement markings are designed for use as durable legend rolls on exterior pavement surfaces. The material provides a non-transparent, skid-resistant finish in white, suitable for professional traffic marking applications.

  • Meets AASHTO M-249, AASHTO T-250, ASTM D 4592, ASTM D 6628, ASTM E 1710, and ASTM E 303 standards
  • Application method requires a hand torch with an application temperature range of 32°F to 275°F
  • Durable construction with 90 mil thickness
  • Fast dry time of 2 min
  • Shelf life of 2 yr
  • Dimensions of 30 ft Lg by 16 in Wd

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.