Learn More
Grainger RAE Pavement Pre-Determined Message, Alkyd Thermoplastic, White, 1 ea
Catalog No. 502784894
Encompass_Preferred
This preformed thermoplastic pavement marking is designed for school zone identification and messaging. The material is supplied as a ready-to-use, pre-cut message that adheres to pavement surfaces via heat application.
  • Composed of durable alkyd thermoplastic for long-term performance.
  • Embedded with high-quality reflective glass beads for visibility.
  • Skid resistant with a minimum initial BPN of 45.
  • Easily installed on pavement surfaces using a hand torch.
  • Environmentally friendly formulation featuring zero VOC content and heavy metal-free pigments.
  • Compliant with MUTCD, ASTM D 6628, ASTM E 303, FHWA, and AASHTO M-249 standards.
  • Ready for traffic exposure within 5 to 10 minutes after cooling.
Catalog No. 50-278-4894 Supplier Grainger Supplier No. 18E556
This preformed thermoplastic pavement marking is designed for school zone identification and messaging. The material is supplied as a ready-to-use, pre-cut message that adheres to pavement surfaces via heat application.
  • Composed of durable alkyd thermoplastic for long-term performance.
  • Embedded with high-quality reflective glass beads for visibility.
  • Skid resistant with a minimum initial BPN of 45.
  • Easily installed on pavement surfaces using a hand torch.
  • Environmentally friendly formulation featuring zero VOC content and heavy metal-free pigments.
  • Compliant with MUTCD, ASTM D 6628, ASTM E 303, FHWA, and AASHTO M-249 standards.
  • Ready for traffic exposure within 5 to 10 minutes after cooling.