Learn More
Grainger Rae Preformed Thermoplastic Pavement Marking, Stop, 8 ft, 6 1/8 ft, 90 mil, 1 ea
Catalog No. 502778011
Encompass_Preferred
These preformed thermoplastic pavement markings provide a durable and reflective solution for traffic signaling. Designed for exterior pavement use, this white Stop legend is constructed from hot applied thermoplastic with a 90 mil thickness to ensure longevity and visibility. The material features enhanced reflectivity through integrated glass beads and offers skid resistance for safety.
  • Reflective glass beads for enhanced visibility
  • Complies with ASTM D 6628, ASTM E 303, MUTCD, FHWA, and AASHTO M-249 standards
  • Easy application using a hand torch
  • Skid resistant surface
  • 2 yr shelf life
  • 0.0 g/l VOC content
Catalog No. 50-277-8011 Supplier Grainger Supplier No. 18E535
These preformed thermoplastic pavement markings provide a durable and reflective solution for traffic signaling. Designed for exterior pavement use, this white Stop legend is constructed from hot applied thermoplastic with a 90 mil thickness to ensure longevity and visibility. The material features enhanced reflectivity through integrated glass beads and offers skid resistance for safety.
  • Reflective glass beads for enhanced visibility
  • Complies with ASTM D 6628, ASTM E 303, MUTCD, FHWA, and AASHTO M-249 standards
  • Easy application using a hand torch
  • Skid resistant surface
  • 2 yr shelf life
  • 0.0 g/l VOC content