Learn More
Grainger RAE Pavement Pre-Determined Message, X-Ing, White, 8 ft Lg, 6-3/4 ft Wd, 1 ea
Catalog No. 502778018
Encompass_Preferred
These preformed thermoplastic pavement markings are designed for enhanced road safety and visibility. This durable X-Ing symbol is engineered for exterior use on pavement and features premium glass beads embedded throughout the material for superior day and night visibility.
  • Provides high-visibility traffic markings
  • Compatible with standard hand torch application
  • Includes embedded reflective glass beads
  • Compliant with ASTM D 6628, ASTM E 303, MUTCD, FHWA, and AASHTO M-249 standards
  • Constructed from 90 mil thick thermoplastic material
  • Designed for long-lasting performance with a 2 yr shelf life
Catalog No. 50-277-8018 Supplier Grainger Supplier No. 18E538
These preformed thermoplastic pavement markings are designed for enhanced road safety and visibility. This durable X-Ing symbol is engineered for exterior use on pavement and features premium glass beads embedded throughout the material for superior day and night visibility.
  • Provides high-visibility traffic markings
  • Compatible with standard hand torch application
  • Includes embedded reflective glass beads
  • Compliant with ASTM D 6628, ASTM E 303, MUTCD, FHWA, and AASHTO M-249 standards
  • Constructed from 90 mil thick thermoplastic material
  • Designed for long-lasting performance with a 2 yr shelf life