Learn More
American Radiolabeled Chemicals Inc PREG-3A-3H 25UC
SDP
Catalog No. 50901227
Encompass
Pregnan-3a-ol-20-one, 5a-9,11,12-3H(N) 25uCi in ethanol, 1 mCi/ml, S.A. 40-70 Ci/mmol, M.W. 318, Shipped in Dry Ice, Exclusive
Catalog No. 50-901-227 Supplier American Radiolabeled Chemicals Inc Supplier No. ART 180825UC
Pregnan-3a-ol-20-one, 5a-9,11,12-3H(N) 25uCi in ethanol, 1 mCi/ml, S.A. 40-70 Ci/mmol, M.W. 318, Shipped in Dry Ice, Exclusive